Tisk…

4,5-Difluoro-2-nitrotoluene 98%

Dodavatel: Apollo Scientific
Nebezpečí

PC4368-100G PC4368-25G PC4368-5G
APOSPC4368-100GEA 4960 CZK
APOSPC4368-100G APOSPC4368-25G APOSPC4368-5G
4,5-Difluoro-2-nitrotoluene 98%
4,5-Difluoro-2-nitrotoluene
Vzorec: C₇H₅F₂NO₂
Storage Temperature: Ambient
MDL Number: MFCD04973772
CAS číslo: 127371-50-0
UN: 2810
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR