Tisk…

tert-Butyl-4-(2,2,2-trifluoroacetamido)piperazine-1-carboxylate

Dodavatel: Apollo Scientific

PC430340-1G
APOSPC430340-1GEA 15540 CZK
APOSPC430340-1G
tert-Butyl-4-(2,2,2-trifluoroacetamido)piperazine-1-carboxylate
tert-Butyl-4-(2,2,2-trifluoroacetamido)piperazine-1-carboxylate
Vzorec: C₁₁H₁₈F₃N₃O₃
Storage Temperature: Ambient
MDL Number: MFCD12912661
CAS číslo: 1198286-22-4

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR