Tisk…

4-Bromo-2-chloro-3-fluorophenol 95+%

Dodavatel: Apollo Scientific

PC430005-250MG PC430005-1G
APOSPC430005-250MGEA 1700 CZK
APOSPC430005-250MG APOSPC430005-1G
4-Bromo-2-chloro-3-fluorophenol 95+%
4-Bromo-2-chloro-3-fluorophenol
Vzorec: C₆H₃BrClFO
Storage Temperature: Ambient
MDL Number: MFCD11849932
CAS číslo: 1233026-52-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR