Tisk…

4,4-Difluorocyclohexanecarboxaldehyde 97%

Dodavatel: Apollo Scientific

PC421133-1G PC421133-500MG PC421133-5G
APOSPC421133-1GEA 12040 CZK
APOSPC421133-1G APOSPC421133-500MG APOSPC421133-5G
4,4-Difluorocyclohexanecarboxaldehyde 97%
4,4-Difluorocyclohexanecarboxaldehyde
Vzorec: C₇H₁₀F₂O
Bod varu: 55…57 °C/6mmHg
Storage Temperature: Freezer
MDL Number: MFCD09263468
CAS číslo: 265108-36-9
UN: 1989
ADR: 3,III

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR