Tisk…

Sodium 3-fluoro-2-oxopropanoate monohydrate 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: 3-Fluoro-2-oxopropanoic acid sodium salt monohydrate, 3-Fluoropyruvic acid sodium salt monohydrate

PC4205-1G PC4205-5G
APOSPC4205-1GEA 12370 CZK
APOSPC4205-1G APOSPC4205-5G
Sodium 3-fluoro-2-oxopropanoate monohydrate 98%
Sodium 3-fluoro-2-oxopropanoate monohydrate
Vzorec: C₃H₂FO₃·Na·H₂O
Bod tání: 145 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00149108
CAS číslo: 238754-68-2
EINECS: 220-880-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR