Tisk…

2-Fluorophenyl isocyanate 98%

Dodavatel: Apollo Scientific
Nebezpečí

PC4164-25G PC4164-5G PC4164-1G
APOSPC4164-25GEA 3840 CZK
APOSPC4164-25G APOSPC4164-5G APOSPC4164-1G
2-Fluorophenyl isocyanate 98%
2-Fluorophenyl isocyanate

  • May form small amount of sediment.

Vzorec: C₇H₄FNO
Bod varu: 56…58 °C/10mmHg
Hustota: 1,222 g/cm³ (25 °C)
Bod vzplanutí: 50 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001996
CAS číslo: 16744-98-2
EINECS: 240-804-0
UN: 3080
ADR: 6.1+3,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR