Tisk…

2-Fluorophenylhydrazinium chloride 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: (2-Fluorophenyl)hydrazine hydrochloride, (2-Fluorphenyl)hydrazin HCl, 1-(2-Fluorophenyl)hydrazine hydrochloride, 2-Fluorophenylhydrazine hydrochloride

PC4152-100G PC4152-25G
APOSPC4152-100GEA 2150 CZK
APOSPC4152-100G APOSPC4152-25G
2-Fluorophenylhydrazinium chloride 98%
2-Fluorophenylhydrazinium chloride
Vzorec: C₆H₈ClFN₂
MW: 162.59
Bod tání: 200…205 °C (rozkl.)
Storage Temperature: Refrigerator
MDL Number: MFCD00012927
CAS číslo: 2924-15-4
EINECS: 220-885-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR