Tisk…

2-Ethoxy-6-trifluoromethylpyridine-4-boronic acid, pinacol ester 96%

Dodavatel: Apollo Scientific

PC412398-1G PC412398-5G
APOSPC412398-1GEA 7250 CZK
APOSPC412398-1G APOSPC412398-5G
2-Ethoxy-6-trifluoromethylpyridine-4-boronic acid, pinacol ester 96%
2-Ethoxy-6-trifluoromethylpyridine-4-boronic acid, pinacol ester
Vzorec: C₁₄H₁₉BF₃NO₃
Storage Temperature: Ambient
MDL Number: MFCD17214287
CAS číslo: 1256359-30-4

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR