Tisk…

5-Amino-2-fluorobenzeneboronic acid pinacol ester 95+%

Dodavatel: Apollo Scientific

PC412222-5G PC412222-1G
APOSPC412222-5GEA 19910 CZK
APOSPC412222-5G APOSPC412222-1G
5-Amino-2-fluorobenzeneboronic acid pinacol ester 95+%
5-Amino-2-fluorobenzeneboronic acid pinacol ester
Vzorec: C₁₂H₁₇BFNO₂
Storage Temperature: Ambient
MDL Number: MFCD16996293
CAS číslo: 1152441-29-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR