Tisk…

1-{[5-Methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]methyl}-1H-pyrazole-3-carboxylic acid

Dodavatel: Apollo Scientific

PC410458-1G
APOSPC410458-1GEA 14680 CZK
APOSPC410458-1G
1-{[5-Methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]methyl}-1H-pyrazole-3-carboxylic acid
1-{[5-Methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]methyl}-1H-pyrazole-3-carboxylic acid
Vzorec: C₁₀H₉F₃N₄O₂
Storage Temperature: Ambient
MDL Number: MFCD04967761
CAS číslo: 1006459-10-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR