Tisk…

1-(2,3-Difluorophenyl)-N-[(1,5-dimethyl-1H-pyrazol-4-yl)methyl]methanamine

Dodavatel: Apollo Scientific

PC410437-1G
APOSPC410437-1GEA 23580 CZK
APOSPC410437-1G
1-(2,3-Difluorophenyl)-N-[(1,5-dimethyl-1H-pyrazol-4-yl)methyl]methanamine
1-(2,3-Difluorophenyl)-N-[(1,5-dimethyl-1H-pyrazol-4-yl)methyl]methanamine
Vzorec: C₁₃H₁₅F₂N₃
Storage Temperature: Ambient
MDL Number: MFCD08696832
CAS číslo: 1006437-26-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR