Tisk…

2-({4-Ethyl-5-[1-methyl-3-(trifluoromethyl)-1H-pyrazol-5-yl]-4H-1,2,4-triazol-3-yl}sulphanyl)acetohydrazide

Dodavatel: Apollo Scientific

PC410365-1G
APOSPC410365-1GEA 14680 CZK
APOSPC410365-1G
2-({4-Ethyl-5-[1-methyl-3-(trifluoromethyl)-1H-pyrazol-5-yl]-4H-1,2,4-triazol-3-yl}sulphanyl)acetohydrazide
2-({4-Ethyl-5-[1-methyl-3-(trifluoromethyl)-1H-pyrazol-5-yl]-4H-1,2,4-triazol-3-yl}sulphanyl)acetohydrazide
Vzorec: C₁₁H₁₄F₃N₇OS
MW: 349,335 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03421859
CAS číslo: 1005700-45-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR