Tisk…

N-Piperidin-4-yl-2-(trifluoromethyl)-1,6-naphthyridin-5-amine hydrochloride monohydrate 95+%

Dodavatel: Apollo Scientific

PC409506-1G PC409506-10G PC409506-5G
APOSPC409506-1GEA 18620 CZK
APOSPC409506-1G APOSPC409506-10G APOSPC409506-5G
N-Piperidin-4-yl-2-(trifluoromethyl)-1,6-naphthyridin-5-amine hydrochloride monohydrate 95+%
N-Piperidin-4-yl-2-(trifluoromethyl)-1,6-naphthyridin-5-amine hydrochloride monohydrate
Vzorec: C₁₄H₁₈ClF₃N₄O
Storage Temperature: Ambient
MDL Number: MFCD08567943
CAS číslo: 1452488-43-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR