Tisk…

[1-(3-Chloro-5-trifluoromethyl-pyridin-2-yl)-pyrrolidin-3-yl]-carbamic acid tert-butyl ester

Dodavatel: Apollo Scientific

PC408387-500MG
APOSPC408387-500MGEA 26920 CZK
APOSPC408387-500MG
[1-(3-Chloro-5-trifluoromethyl-pyridin-2-yl)-pyrrolidin-3-yl]-carbamic acid tert-butyl ester
[1-(3-Chloro-5-trifluoromethyl-pyridin-2-yl)-pyrrolidin-3-yl]-carbamic acid tert-butyl ester
Vzorec: C₁₅H₁₉ClF₃N₃O₂
Storage Temperature: Ambient
MDL Number: MFCD20921630
CAS číslo: 1354448-68-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR