Tisk…

(R)-3-[1-(4-Fluoro-benzyl)-1H-benzoimidazol-2-yloxy]-pyrrolidine-1-carboxylic acid tert-butyl ester

Dodavatel: Apollo Scientific

PC408382-500MG
APOSPC408382-500MGEA 17630 CZK
APOSPC408382-500MG
(R)-3-[1-(4-Fluoro-benzyl)-1H-benzoimidazol-2-yloxy]-pyrrolidine-1-carboxylic acid tert-butyl ester
(R)-3-[1-(4-Fluoro-benzyl)-1H-benzoimidazol-2-yloxy]-pyrrolidine-1-carboxylic acid tert-butyl ester
Vzorec: C₂₃H₂₆FN₃O₃
Storage Temperature: Ambient
MDL Number: MFCD21099002
CAS číslo: 1354006-88-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR