Tisk…

2,2,3,4,4,4-Hexafluorobutyric acid 97%

Dodavatel: Apollo Scientific
Nebezpečí

PC408093-1G PC408093-5G
APOSPC408093-1GEA 4900 CZK
APOSPC408093-1G APOSPC408093-5G
2,2,3,4,4,4-Hexafluorobutyric acid 97%
2,2,3,4,4,4-Hexafluorobutyric acid
Vzorec: C₄H₂F₆O₂
Bod varu: 140 °C (740 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD16621311
CAS číslo: 379-90-8
UN: 3265
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR