Tisk…

(Nonafluorobutyl)undecafluorocyclohexane

Dodavatel: Apollo Scientific
Varování

PC408080-25G PC408080-5G
APOSPC408080-25GEA 1460 CZK
APOSPC408080-25G APOSPC408080-5G
(Nonafluorobutyl)undecafluorocyclohexane
(Nonafluorobutyl)undecafluorocyclohexane
Vzorec: C₁₀F₂₀
Bod varu: 145…147 °C
Hustota: 1,899 g/cm³ (25 °C)
Storage Temperature: Ambient
CAS číslo: 374-60-7
EINECS: 206-777-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR