Tisk…

2,4-Difluoro-6-hydroxybenzoic acid methyl ester 98%

Dodavatel: Apollo Scientific

PC408069-1G PC408069-5G
APOSPC408069-1GEA 2370 CZK
APOSPC408069-1G APOSPC408069-5G
2,4-Difluoro-6-hydroxybenzoic acid methyl ester 98%
2,4-Difluoro-6-hydroxybenzoic acid methyl ester
Vzorec: C₈H₆F₂O₃
Storage Temperature: Ambient
MDL Number: MFCD06203802
CAS číslo: 773874-16-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR