Tisk…

3-Chloro-5-(pentafluorosulphur)benzoic acid 97%

Dodavatel: Apollo Scientific

PC405677-1G PC405677-5G
APOSPC405677-1GEA 12010 CZK
APOSPC405677-1G APOSPC405677-5G
3-Chloro-5-(pentafluorosulphur)benzoic acid 97%
3-Chloro-5-(pentafluorosulphur)benzoic acid
Vzorec: C₇H₄ClF₅O₂S
Storage Temperature: Ambient
MDL Number: MFCD16652415
CAS číslo: 1448317-67-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR