Tisk…

2-Chloro-7-fluoroquinoline-3-carboxaldehyde

Dodavatel: Apollo Scientific
Varování

PC404559-250MG PC404559-1G
APOSPC404559-250MGEA 4140 CZK
APOSPC404559-250MG APOSPC404559-1G
2-Chloro-7-fluoroquinoline-3-carboxaldehyde
2-Chloro-7-fluoroquinoline-3-carboxaldehyde
Vzorec: C₁₀H₅ClFNO
Storage Temperature: Ambient
MDL Number: MFCD09787681
CAS číslo: 745830-16-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR