Tisk…

Methyl trifluoroacrylate

Dodavatel: Apollo Scientific
Varování

PC4032-25G PC4032-1G PC4032-5G
APOSPC4032-25GEA 18220 CZK
APOSPC4032-25G APOSPC4032-1G APOSPC4032-5G
Methyl trifluoroacrylate
Methyl trifluoroacrylate
Vzorec: C₄H₃F₃O₂
Bod varu: 85…86 °C
Hustota: 1,3259 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00077566
CAS číslo: 392-41-6
EINECS: 206-875-7

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR