Tisk…

3-(4-(Trifluoromethyl)phenyl)piperidin-3-ol hydrochloride

Dodavatel: Apollo Scientific

PC402144-1G PC402144-250MG
APOSPC402144-1GEA 18190 CZK
APOSPC402144-1G APOSPC402144-250MG
3-(4-(Trifluoromethyl)phenyl)piperidin-3-ol hydrochloride
3-(4-(Trifluoromethyl)phenyl)piperidin-3-ol hydrochloride
Vzorec: C₁₂H₁₄F₃NO·ClH
Storage Temperature: Refrigerator
MDL Number: MFCD28108755
CAS číslo: 2409589-75-7

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR