Tisk…

1-(Azetidin-3-yl)-4-fluoropiperidine hydrochloride

Dodavatel: Apollo Scientific

PC401504-1G
APOSPC401504-1GEA 36370 CZK
APOSPC401504-1G
1-(Azetidin-3-yl)-4-fluoropiperidine hydrochloride
1-(Azetidin-3-yl)-4-fluoropiperidine hydrochloride
Vzorec: C₈H₁₅FN₂·ClH
Storage Temperature: Ambient
MDL Number: MFCD25460336
CAS číslo: 1449117-63-8

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR