Tisk…

2-Chloro-5-trifluoromethyl[1,3,4]thiadiazole 95+%

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Chloro-5-trifluoromethyl-[1,3,4]-thiadiazole

PC401017-5G
APOSPC401017-5GEA 12370 CZK
APOSPC401017-5G
2-Chloro-5-trifluoromethyl[1,3,4]thiadiazole 95+%
2-Chloro-5-trifluoromethyl[1,3,4]thiadiazole
Vzorec: C₃ClF₃N₂S
MW: 188.56 g/mol
Bod varu: 64…66 °C/20mmHg
Storage Temperature: Ambient
MDL Number: MFCD04972766
CAS číslo: 53645-98-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR