Tisk…

Quinolinium fluorochromate (QFC) on alumina

Dodavatel: Apollo Scientific
Varování

PC3876-25G PC3876-5G
APOSPC3876-25GEA 14090 CZK
APOSPC3876-25G APOSPC3876-5G
Quinolinium fluorochromate (QFC) on alumina
Quinolinium fluorochromate (QFC)

Selectively oxidises aliphatic primary and secondary alcohols and acyclic alcohols to their corresponding carbonyl compounds in good yield, ask for reference. Minimum activity 1mmol QFC per gram of alumina

Vzorec: C₉H₇N·CrFHO₃
MW: 249.159 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02093330
CAS číslo: 143440-93-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR