Tisk…

tert-Butyl pentafluoropropionate 98%

Dodavatel: Apollo Scientific

PC3772-25G PC3772-5G
APOSPC3772-25GEA 18880 CZK
APOSPC3772-25G APOSPC3772-5G
tert-Butyl pentafluoropropionate 98%
tert-Butyl pentafluoropropionate
Vzorec: C₇H₉F₅O₂
Storage Temperature: Ambient
MDL Number: MFCD00464380
CAS číslo: 55258-28-1

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR