Tisk…

Ethyl-2-methyl-6-(trifluoromethyl)nicotinate 95+%

Dodavatel: Apollo Scientific

PC3721-10G PC3721-5G PC3721-1G PC3721-250MG
APOSPC3721-10GEA 0 CZK
APOSPC3721-10G APOSPC3721-5G APOSPC3721-1G APOSPC3721-250MG
Ethyl-2-methyl-6-(trifluoromethyl)nicotinate 95+%
Ethyl-2-methyl-6-(trifluoromethyl)nicotinate
Vzorec: C₁₀H₁₀F₃NO₂
MW: 233.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10688283
CAS číslo: 380355-65-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR