Tisk…

2,2,3,3-Tetrafluoropropyltriethylammoniumtriflate 97%

Dodavatel: Apollo Scientific
Varování

PC3382-25G PC3382-5G
APOSPC3382-25GEA 12340 CZK
APOSPC3382-25G APOSPC3382-5G
2,2,3,3-Tetrafluoropropyltriethylammoniumtriflate 97%
2,2,3,3-Tetrafluoropropyltriethylammoniumtriflate
Vzorec: C₁₀H₁₈F₇NO₃S
MW: 365.31 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04038359
CAS číslo: 1330184-85-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR