Tisk…

Perfluorbutanesulphonylamide(N-methyl)acetate 97%

Dodavatel: Apollo Scientific
Varování

PC32961-100MG PC32961-250MG
APOSPC32961-100MGEA 11540 CZK
APOSPC32961-100MG APOSPC32961-250MG
Perfluorbutanesulphonylamide(N-methyl)acetate 97%
Perfluorbutanesulphonylamide(N-methyl)acetate
Vzorec: C₇H₆F₉NO₄S
Storage Temperature: Ambient
MDL Number: MFCD32011191
CAS číslo: 159381-10-9

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR