Tisk…

3-(3-Fluorophenyl)acryloyl chloride

Dodavatel: Apollo Scientific

PC32865-1G PC32865-250MG PC32865-500MG
APOSPC32865-1GEA 13790 CZK
APOSPC32865-1G APOSPC32865-250MG APOSPC32865-500MG
3-(3-Fluorophenyl)acryloyl chloride
3-(3-Fluorophenyl)acryloyl chloride
Vzorec: C₉H₆ClFO
Storage Temperature: Ambient
MDL Number: MFCD03424739
CAS číslo: 39098-87-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR