Tisk…

5-Chloro-3-(trifluoromethyl)-1H-pyrazole-4-carbaldehyde

Dodavatel: Apollo Scientific

PC32853-1G PC32853-250MG PC32853-500MG
APOSPC32853-1GEA 31610 CZK
APOSPC32853-1G APOSPC32853-250MG APOSPC32853-500MG
5-Chloro-3-(trifluoromethyl)-1H-pyrazole-4-carbaldehyde
5-Chloro-3-(trifluoromethyl)-1H-pyrazole-4-carbaldehyde
Vzorec: C₅H₂ClF₃N₂O
Storage Temperature: Ambient
MDL Number: MFCD07375187
CAS číslo: 154357-44-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR