Tisk…

4-Fluoro-2-methyl-5-nitroanisole

Dodavatel: Apollo Scientific
Varování

Synonyms: 5-Fluoro-2-methoxy-4-nitrotoluene, 4-Fluoro-2-methyl-5-nitrophenyl methyl ether, 1-Fluoro-4-methoxy-5-methyl-2-nitrobenzene

PC32802-1G PC32802-250MG PC32802-5G
APOSPC32802-1GEA 3870 CZK
APOSPC32802-1G APOSPC32802-250MG APOSPC32802-5G
4-Fluoro-2-methyl-5-nitroanisole
4-Fluoro-2-methyl-5-nitroanisole
Vzorec: C₈H₈FNO₃
MW: 185.152 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27937156
CAS číslo: 134882-63-6

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR