Tisk…

3-Fluoro-5-iodobenzyl alcohol

Dodavatel: Apollo Scientific
Varování

PC32750-1G PC32750-250MG PC32750-5G
APOSPC32750-1GEA 2320 CZK
APOSPC32750-1G APOSPC32750-250MG APOSPC32750-5G
3-Fluoro-5-iodobenzyl alcohol
3-Fluoro-5-iodobenzyl alcohol
Vzorec: C₇H₆FIO
Storage Temperature: Ambient
MDL Number: MFCD18396948
CAS číslo: 1261837-87-9

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR