Tisk…

2,5-Dichloro-N'-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]pyridine-3-carbohydrazide

Dodavatel: Apollo Scientific
Varování

PC32676-100MG
APOSPC32676-100MGEA 2070 CZK
APOSPC32676-100MG
2,5-Dichloro-N'-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]pyridine-3-carbohydrazide
2,5-Dichloro-N'-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]pyridine-3-carbohydrazide
Vzorec: C₁₂H₆Cl₃F₃N₄O
Storage Temperature: Ambient
MDL Number: MFCD00833282
CAS číslo: 680217-58-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR