Tisk…

3-Bromo-5-fluoroanthranilic acid

Dodavatel: Apollo Scientific
Varování

PC32490-1G PC32490-5G PC32490-10G
APOSPC32490-1GEA 622 CZK
APOSPC32490-1G APOSPC32490-5G APOSPC32490-10G
3-Bromo-5-fluoroanthranilic acid
3-Bromo-5-fluoroanthranilic acid
Vzorec: C₇H₅BrFNO₂
Storage Temperature: Ambient
MDL Number: MFCD00662509
CAS číslo: 259269-84-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR