Tisk…

(4,5-Dichloro-1H-imidazol-1-yl)methyl [4-(trifluoromethoxy)phenyl]carbamate

Dodavatel: Apollo Scientific
Varování

Synonyms: 4,5-Dichloro-1-[({[4-(trifluoromethoxy)phenyl]carbamoyl}oxy)methyl]-1H-imidazole

PC31829-250MG
APOSPC31829-250MGEA 1990 CZK
APOSPC31829-250MG
(4,5-Dichloro-1H-imidazol-1-yl)methyl [4-(trifluoromethoxy)phenyl]carbamate
(4,5-Dichloro-1H-imidazol-1-yl)methyl [4-(trifluoromethoxy)phenyl]carbamate
Vzorec: C₁₂H₈Cl₂F₃N₃O₃
MW: 370.111 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00110749
CAS číslo: 650592-39-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR