Tisk…

5-Bromo-2-methyl-3-(trifluoromethyl)phenylacetic acid 98%

Dodavatel: Apollo Scientific

PC303590-1G PC303590-5G
APOSPC303590-1GEA 10350 CZK
APOSPC303590-1G APOSPC303590-5G
5-Bromo-2-methyl-3-(trifluoromethyl)phenylacetic acid 98%
5-Bromo-2-methyl-3-(trifluoromethyl)phenylacetic acid
Vzorec: C₁₀H₈BrF₃O₂
Storage Temperature: Ambient
MDL Number: MFCD32640165
CAS číslo: 2383520-58-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR