Tisk…

3-Bromo-4-chloro-5-(trifluoromethyl)benzyl bromide

Dodavatel: Apollo Scientific

PC303565-1G PC303565-5G
APOSPC303565-1GEA 7940 CZK
APOSPC303565-1G APOSPC303565-5G
3-Bromo-4-chloro-5-(trifluoromethyl)benzyl bromide
3-Bromo-4-chloro-5-(trifluoromethyl)benzyl bromide
Vzorec: C₈H₄Br₂ClF₃
Storage Temperature: Ambient
MDL Number: MFCD32640143
CAS číslo: 2386636-01-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR