Tisk…

4-Phenoxy-2-(trifluoromethyl)benzoic acid 97%

Dodavatel: Apollo Scientific

PC303551-1G
APOSPC303551-1GEA 8270 CZK
APOSPC303551-1G
4-Phenoxy-2-(trifluoromethyl)benzoic acid 97%
4-Phenoxy-2-(trifluoromethyl)benzoic acid
Vzorec: C₁₄H₉F₃O₃
Storage Temperature: Ambient
MDL Number: MFCD32189207
CAS číslo: 1253188-35-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR