Tisk…

2-Bromo-4,6-bis(trifluoromethyl)phenylacetic acid

Dodavatel: Apollo Scientific

PC303532-1G PC303532-5G
APOSPC303532-1GEA 0 CZK
APOSPC303532-1G APOSPC303532-5G
2-Bromo-4,6-bis(trifluoromethyl)phenylacetic acid
2-Bromo-4,6-bis(trifluoromethyl)phenylacetic acid
Vzorec: C₁₀H₅BrF₆O₂
Storage Temperature: Ambient
MDL Number: MFCD28728131
CAS číslo: 1807217-12-4

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR