Tisk…

2-Bromo-4,6-bis(trifluoromethyl)phenylacetonitrile

Dodavatel: Apollo Scientific

PC303531-1G PC303531-5G
APOSPC303531-1GEA 11250 CZK
APOSPC303531-1G APOSPC303531-5G
2-Bromo-4,6-bis(trifluoromethyl)phenylacetonitrile
2-Bromo-4,6-bis(trifluoromethyl)phenylacetonitrile
Vzorec: C₁₀H₄BrF₆N
Storage Temperature: Ambient
MDL Number: MFCD28728179
CAS číslo: 1805277-63-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR