Tisk…

2-Bromo-4,6-bis(trifluoromethyl)benzaldehyde 97%

Dodavatel: Apollo Scientific

PC303528-1G
APOSPC303528-1GEA 7940 CZK
APOSPC303528-1G
2-Bromo-4,6-bis(trifluoromethyl)benzaldehyde 97%
2-Bromo-4,6-bis(trifluoromethyl)benzaldehyde
Vzorec: C₉H₃BrF₆O
Storage Temperature: Ambient
MDL Number: MFCD28727819
CAS číslo: 1415130-39-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR