Tisk…

3-Bromo-2-fluoro-5-(trifluoromethyl)benzylamine

Dodavatel: Apollo Scientific

PC303469-1G PC303469-5G
APOSPC303469-1GEA 11910 CZK
APOSPC303469-1G APOSPC303469-5G
3-Bromo-2-fluoro-5-(trifluoromethyl)benzylamine
3-Bromo-2-fluoro-5-(trifluoromethyl)benzylamine
Vzorec: C₈H₆BrF₄N
Storage Temperature: Ambient
MDL Number: MFCD20485012
CAS číslo: 871325-57-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR