Tisk…

3-Bromo-2-fluoro-5-(trifluoromethyl)benzyl bromide 97%

Dodavatel: Apollo Scientific

PC303466-1G PC303466-5G PC303466-25G
APOSPC303466-1GEA 7940 CZK
APOSPC303466-1G APOSPC303466-5G APOSPC303466-25G
3-Bromo-2-fluoro-5-(trifluoromethyl)benzyl bromide 97%
3-Bromo-2-fluoro-5-(trifluoromethyl)benzyl bromide
Vzorec: C₈H₄Br₂F₄
Storage Temperature: Ambient
MDL Number: MFCD32265218
CAS číslo: 2092564-82-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR