Tisk…

5-Bromo-3-chloro-2-fluorobenzylamine

Dodavatel: Apollo Scientific

PC303463-1G PC303463-250MG
APOSPC303463-1GEA 19010 CZK
APOSPC303463-1G APOSPC303463-250MG
5-Bromo-3-chloro-2-fluorobenzylamine
5-Bromo-3-chloro-2-fluorobenzylamine
Vzorec: C₇H₆BrClFN
MW: 238.485 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23163937
CAS číslo: 1386459-84-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR