Tisk…

4,6-Dichloro-2-fluoro-3-methoxybenzyl alcohol 97%

Dodavatel: Apollo Scientific

PC303368-1G PC303368-5G
APOSPC303368-1GEA 7870 CZK
APOSPC303368-1G APOSPC303368-5G
4,6-Dichloro-2-fluoro-3-methoxybenzyl alcohol 97%
4,6-Dichloro-2-fluoro-3-methoxybenzyl alcohol
Vzorec: C₈H₇Cl₂FO₂
Storage Temperature: Ambient
MDL Number: MFCD30726937
CAS číslo: 2149598-95-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR