Tisk…

2-Bromo-5-chloro-4-(trifluoromethoxy)aniline 97%

Dodavatel: Apollo Scientific

PC303350-5G PC303350-1G
APOSPC303350-5GEA 22450 CZK
APOSPC303350-5G APOSPC303350-1G
2-Bromo-5-chloro-4-(trifluoromethoxy)aniline 97%
2-Bromo-5-chloro-4-(trifluoromethoxy)aniline
Vzorec: C₇H₄BrClF₃NO
Storage Temperature: Ambient
MDL Number: MFCD28734569
CAS číslo: 868406-81-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR