Tisk…

2-Chloro-4-fluoro-5-methylphenylacetic acid 98%

Dodavatel: Apollo Scientific

PC303341-1G
APOSPC303341-1GEA 11180 CZK
APOSPC303341-1G
2-Chloro-4-fluoro-5-methylphenylacetic acid 98%
2-Chloro-4-fluoro-5-methylphenylacetic acid
Vzorec: C₉H₈ClFO₂
MW: 202,61 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09925052
CAS číslo: 1000522-29-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR