Tisk…

3,5-Bis(trifluoromethyl)-α-methylbenzyl bromide 98%

Dodavatel: Apollo Scientific

PC303333-5G PC303333-1G
APOSPC303333-5GEA 22680 CZK
APOSPC303333-5G APOSPC303333-1G
3,5-Bis(trifluoromethyl)-α-methylbenzyl bromide 98%
3,5-Bis(trifluoromethyl)-α-methylbenzyl bromide
Vzorec: C₁₀H₇BrF₆
Storage Temperature: Ambient
MDL Number: MFCD18762083
CAS číslo: 160376-84-1
UN: 3265
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR