Tisk…

4-Chloro-2-fluoro-5-methylbenzenesulphonyl chloride

Dodavatel: Apollo Scientific

PC303329-1G
APOSPC303329-1GEA 12800 CZK
APOSPC303329-1G
4-Chloro-2-fluoro-5-methylbenzenesulphonyl chloride
4-Chloro-2-fluoro-5-methylbenzenesulphonyl chloride
Vzorec: C₇H₅Cl₂FO₂S
Storage Temperature: Ambient
MDL Number: MFCD08060548
CAS číslo: 868755-70-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR